Learn More
MSC Vermont Gage Pin Gage, 0.1249 Dia, 2 Inch Length, Class X Plus, 4 Micro
Catalog No. 5000529335
Encompass
This pin gage is designed for precise measurement applications, suitable for calibrating measuring equipment or directly gaging parts. It features a specific diameter and tight tolerance, ensuring accuracy, and is constructed from durable tool steel.
  • Diameter of 0.1249 inch
  • Tolerance of 0.00004 inch
  • Constructed from tool steel
  • Ideal for calibration of measuring equipment
  • Can be used to gage parts directly
  • Includes certificate of accuracy with NIST traceability
  • Length of 2 inches
  • 4 micro coated finish
Catalog No. 50-005-29335 Supplier MSC Supplier No. 08506727
This pin gage is designed for precise measurement applications, suitable for calibrating measuring equipment or directly gaging parts. It features a specific diameter and tight tolerance, ensuring accuracy, and is constructed from durable tool steel.
  • Diameter of 0.1249 inch
  • Tolerance of 0.00004 inch
  • Constructed from tool steel
  • Ideal for calibration of measuring equipment
  • Can be used to gage parts directly
  • Includes certificate of accuracy with NIST traceability
  • Length of 2 inches
  • 4 micro coated finish