Learn More
MSC Vermont Gage Pin Gage, 0.133 in, Black Guard, Class ZZ, NoGo
Catalog No. 5000529878
Encompass
Vermont Gage pin gages are designed for quick and accurate measurements. They are used to measure hole sizes, calculate distances between holes, gage slot widths, check locations, and for Go/NoGo gaging. These gages can also be used to size micrometers and snap gages.
  • Tool steel construction with 60/65 Rc hardness.
  • 2 in long.
  • Features a 10 microinch finish or better.
  • Round within 0.0001 in.
  • Marked with size and direction of tolerance on gages over 0.0605 in.
  • NoGo (minus) tolerance direction.
Catalog No. 50-005-29878 Supplier MSC Supplier No. 02658268
Vermont Gage pin gages are designed for quick and accurate measurements. They are used to measure hole sizes, calculate distances between holes, gage slot widths, check locations, and for Go/NoGo gaging. These gages can also be used to size micrometers and snap gages.
  • Tool steel construction with 60/65 Rc hardness.
  • 2 in long.
  • Features a 10 microinch finish or better.
  • Round within 0.0001 in.
  • Marked with size and direction of tolerance on gages over 0.0605 in.
  • NoGo (minus) tolerance direction.