Learn More
MSC Vermont Gage Pin Gage, 0.1395 in Diameter, Class ZZ Plus, 2.0000 in OAL
Catalog No. 5000529174
Encompass
This pin gage is designed for precise measurements in various applications including measuring hole sizes, calculating distances, gaging slot widths, checking locations, and Go/No Go gaging. Made from 52100 tool steel, it ensures durability and accuracy. Each pin is NIST traceable and includes a certificate of accuracy.
  • Size electro etched on each piece.
  • Class ZZ Plus tolerance of -0.0000 in/+0.0002 in.
  • All members are 2 in long and centerless lapped.
  • Heat treated to Rc 60 - 62 hardness with a 10 micro finish or better.
  • Measures hole sizes, calculates distances between holes, and gages slot widths.
  • Checks locations and performs Go/No Go gaging.
  • Ideal for quick, accurate, frequent, and varied measurements in micrometers and snap gages.
Catalog No. 50-005-29174 Supplier MSC Supplier No. 03425287
This pin gage is designed for precise measurements in various applications including measuring hole sizes, calculating distances, gaging slot widths, checking locations, and Go/No Go gaging. Made from 52100 tool steel, it ensures durability and accuracy. Each pin is NIST traceable and includes a certificate of accuracy.
  • Size electro etched on each piece.
  • Class ZZ Plus tolerance of -0.0000 in/+0.0002 in.
  • All members are 2 in long and centerless lapped.
  • Heat treated to Rc 60 - 62 hardness with a 10 micro finish or better.
  • Measures hole sizes, calculates distances between holes, and gages slot widths.
  • Checks locations and performs Go/No Go gaging.
  • Ideal for quick, accurate, frequent, and varied measurements in micrometers and snap gages.