Learn More
MSC Vermont Gage Pin Gage, 0.1780 in Diameter, Class X, 2 in Long
Catalog No. 5000532071
Encompass
This Class X pin gage is a precise measurement standard suitable for equipment calibration or direct part gaging. It offers accurate, cost-effective measurement in both shop floor and metrology laboratory applications.
  • Constructed from tool steel for durability.
  • Features a 2 inch long body.
  • Maintains a 4 microinch surface finish or better.
  • Guarantees roundness within 1/2 the diameter tolerance.
  • Offers a diameter tolerance of ±0.000040 "
  • Includes a certificate of accuracy.
  • ISO 17025 accredited and non-accredited calibration certificates are available for purchase.
Catalog No. 50-005-32071 Supplier MSC Supplier No. 88717806
This Class X pin gage is a precise measurement standard suitable for equipment calibration or direct part gaging. It offers accurate, cost-effective measurement in both shop floor and metrology laboratory applications.
  • Constructed from tool steel for durability.
  • Features a 2 inch long body.
  • Maintains a 4 microinch surface finish or better.
  • Guarantees roundness within 1/2 the diameter tolerance.
  • Offers a diameter tolerance of ±0.000040 "
  • Includes a certificate of accuracy.
  • ISO 17025 accredited and non-accredited calibration certificates are available for purchase.