Learn More
MSC Vermont Gage Pin Gage, 0.1815 in Diameter, Class X Minus, Tool Steel, 2 in Long
Catalog No. 5000530738
Encompass
Pin gages are essential tools for quick and accurate measurements, capable of measuring hole sizes, calculating distances between holes, gaging slot widths, and checking locations. They are also suitable for Go/NoGo gaging and sizing in micrometers and snap gages. These gages provide accurate, cost-effective measurement standards for both shop floor and metrology laboratory applications.
  • Class X tolerance for precise measurements
  • NIST traceable for assured accuracy
  • Constructed from tool steel for durability
  • Measures 2 inches in length
  • Features a 4 microinch finish or better
  • Roundness maintained within 1/2 of tolerance
  • Certificate of accuracy included
Catalog No. 50-005-30738 Supplier MSC Supplier No. 88818158
Pin gages are essential tools for quick and accurate measurements, capable of measuring hole sizes, calculating distances between holes, gaging slot widths, and checking locations. They are also suitable for Go/NoGo gaging and sizing in micrometers and snap gages. These gages provide accurate, cost-effective measurement standards for both shop floor and metrology laboratory applications.
  • Class X tolerance for precise measurements
  • NIST traceable for assured accuracy
  • Constructed from tool steel for durability
  • Measures 2 inches in length
  • Features a 4 microinch finish or better
  • Roundness maintained within 1/2 of tolerance
  • Certificate of accuracy included