Learn More
MSC Vermont Gage Pin Gage, 0.2305 inch, Class ZZ, Steel, Minus
Catalog No. 5000531472
Encompass
Vermont Gage Class ZZ pin gages are economical, general-purpose tools designed for various precision measurements. They are commonly used to measure hole sizes, calculate distances between holes, gage slot widths, and check locations. These gages are also suitable for Go/NoGo gaging, offering added wear life compared to gages with closer tolerances.
  • Tolerance of .0002 inch or .005 mm
  • Constructed from tool steel with a 60/65 Rc hardness
  • 2 inch long
  • Features a 10 microinch finish or better
  • Round within .0001 inch or .0025 mm
  • NIST traceable
  • Marked with size and direction of tolerance on gages over .0605 inch
  • Includes a certificate of accuracy
Catalog No. 50-005-31472 Supplier MSC Supplier No. 89023055
Vermont Gage Class ZZ pin gages are economical, general-purpose tools designed for various precision measurements. They are commonly used to measure hole sizes, calculate distances between holes, gage slot widths, and check locations. These gages are also suitable for Go/NoGo gaging, offering added wear life compared to gages with closer tolerances.
  • Tolerance of .0002 inch or .005 mm
  • Constructed from tool steel with a 60/65 Rc hardness
  • 2 inch long
  • Features a 10 microinch finish or better
  • Round within .0001 inch or .0025 mm
  • NIST traceable
  • Marked with size and direction of tolerance on gages over .0605 inch
  • Includes a certificate of accuracy