Learn More
MSC Vermont Gage Pin Gage, 0.2630 inch, Class ZZ Minus, Black Oxide, Steel
Catalog No. 5000529876
Encompass
This 0.2630 inch diameter, Class ZZ Minus pin gage is engineered for precise measurement and inspection. Its black oxide coating provides enhanced durability, corrosion resistance, and visible wear indication for extended service life. Suitable for setting micrometers, verniers, and performing NoGo inspections, it includes a Certificate of Accuracy with NIST traceability.
  • Black oxide finish prevents rust and corrosion, and indicates wear visually.
  • Provides longer useful life.
  • Ideal for setting micrometers, verniers, and other measuring devices.
  • Specifically designed for NoGo inspection.
  • Includes a certificate of accuracy with NIST traceability.
  • Constructed from durable tool steel.
  • Features a 10 microinch finish or better and roundness within 0.0001 "
Catalog No. 50-005-29876 Supplier MSC Supplier No. 02737468
This 0.2630 inch diameter, Class ZZ Minus pin gage is engineered for precise measurement and inspection. Its black oxide coating provides enhanced durability, corrosion resistance, and visible wear indication for extended service life. Suitable for setting micrometers, verniers, and performing NoGo inspections, it includes a Certificate of Accuracy with NIST traceability.
  • Black oxide finish prevents rust and corrosion, and indicates wear visually.
  • Provides longer useful life.
  • Ideal for setting micrometers, verniers, and other measuring devices.
  • Specifically designed for NoGo inspection.
  • Includes a certificate of accuracy with NIST traceability.
  • Constructed from durable tool steel.
  • Features a 10 microinch finish or better and roundness within 0.0001 "