Learn More
MSC Vermont Gage Pin Gage, 0.2660 in Diameter, Class ZZ Minus, Steel
Catalog No. 5000536939
Encompass
This pin gage is a precision measuring tool designed for accurate inspection. Manufactured from steel, it features a 0.2660 in diameter and a 2 in overall length, meeting Class ZZ Minus tolerance for precise measurements. It is suitable for setting micrometers, NoGo inspection, and use with verniers and other measuring devices.
  • Includes certificate of accuracy with NIST traceability
  • Features a 10 micro coating/finish
  • Constructed from durable steel
  • Designed to Class ZZ Minus tolerance
  • 0.2660 in diameter for precise measurement
  • 2 in overall length for versatile application
Catalog No. 50-005-36939 Supplier MSC Supplier No. 89026603
This pin gage is a precision measuring tool designed for accurate inspection. Manufactured from steel, it features a 0.2660 in diameter and a 2 in overall length, meeting Class ZZ Minus tolerance for precise measurements. It is suitable for setting micrometers, NoGo inspection, and use with verniers and other measuring devices.
  • Includes certificate of accuracy with NIST traceability
  • Features a 10 micro coating/finish
  • Constructed from durable steel
  • Designed to Class ZZ Minus tolerance
  • 0.2660 in diameter for precise measurement
  • 2 in overall length for versatile application