Learn More
MSC Vermont Gage Pin Gage, 0.2700 in, Class ZZ Minus, Steel
Catalog No. 5000535797
Encompass
This pin gage is designed for precise measurements of hole sizes. Made from steel, it features a 0.2700 inch diameter and a 2 inch overall length. As a Class ZZ No-Go (Minus) type gage, it is ideal for verifying hole tolerances within +.0000/-.0002 inches. It meets ISO 17025 standards and includes a Certificate of Accuracy with NIST Traceability.
  • Made from steel for durability
  • Measures hole sizes within +.0000/-.0002 in
  • Includes a certificate of accuracy with NIST traceability
  • Has an overall length of 2 in
  • Features a 0.2700 in diameter
  • Class ZZ No-Go type pin gage
  • Uncoated steel finish
  • Tolerance of 0.00020 in
  • Hardness of RC 60/65
  • Meets ISO 17025 standards
  • Cylindrical shape
  • Surface finish of 10 microinch or better
Catalog No. 50-005-35797 Supplier MSC Supplier No. 89027007
This pin gage is designed for precise measurements of hole sizes. Made from steel, it features a 0.2700 inch diameter and a 2 inch overall length. As a Class ZZ No-Go (Minus) type gage, it is ideal for verifying hole tolerances within +.0000/-.0002 inches. It meets ISO 17025 standards and includes a Certificate of Accuracy with NIST Traceability.
  • Made from steel for durability
  • Measures hole sizes within +.0000/-.0002 in
  • Includes a certificate of accuracy with NIST traceability
  • Has an overall length of 2 in
  • Features a 0.2700 in diameter
  • Class ZZ No-Go type pin gage
  • Uncoated steel finish
  • Tolerance of 0.00020 in
  • Hardness of RC 60/65
  • Meets ISO 17025 standards
  • Cylindrical shape
  • Surface finish of 10 microinch or better