Learn More
MSC Vermont Gage Pin Gage, 0.3541 in, Class X, Go, Tool Steel
Catalog No. 5000529885
Encompass
This pin gage is a precision measurement standard designed for calibrating equipment or directly gaging parts. Made from durable tool steel, it features a 0.3541 inch diameter and a 2 inch length. This Class X Go gage is accurate and cost-effective, suitable for both shop floor and metrology laboratory environments.
  • Precision measurement standard for calibration or direct part gaging.
  • Constructed from durable tool steel.
  • 0.3541 inch diameter with a plus tolerance of +0.00004 "
  • 2 inch length.
  • Features a 4 microinch finish or better.
  • Gage roundness maintained within half the diameter tolerance.
  • Includes a certificate of accuracy with NIST traceability.
Catalog No. 50-005-29885 Supplier MSC Supplier No. 03283199
This pin gage is a precision measurement standard designed for calibrating equipment or directly gaging parts. Made from durable tool steel, it features a 0.3541 inch diameter and a 2 inch length. This Class X Go gage is accurate and cost-effective, suitable for both shop floor and metrology laboratory environments.
  • Precision measurement standard for calibration or direct part gaging.
  • Constructed from durable tool steel.
  • 0.3541 inch diameter with a plus tolerance of +0.00004 "
  • 2 inch length.
  • Features a 4 microinch finish or better.
  • Gage roundness maintained within half the diameter tolerance.
  • Includes a certificate of accuracy with NIST traceability.