Learn More
MSC Vermont Gage Pin Gage, 0.3600 in Diameter, 2 in Length, Class X Minus
Catalog No. 5000531963
Encompass
This Vermont Gage Class X Minus Pin Gage is designed with a diameter of 0.3600 inches and a length of 2 inches. It is constructed from Tool Steel with a 4 Micro Coated finish, making it suitable for precise measurement applications and ideal for calibrating equipment or directly gaging parts.
  • Diameter of 0.3600 inches
  • Length of 2 inches
  • Class X Minus for high precision
  • Tolerance of -0.00004 decimal inch
  • Made from durable tool steel
  • Features a 4 micro coated finish
  • Suitable for calibration of measuring equipment
  • Can be used to gage parts directly
  • Includes certificate of accuracy with NIST traceability
Catalog No. 50-005-31963 Supplier MSC Supplier No. 88836002
This Vermont Gage Class X Minus Pin Gage is designed with a diameter of 0.3600 inches and a length of 2 inches. It is constructed from Tool Steel with a 4 Micro Coated finish, making it suitable for precise measurement applications and ideal for calibrating equipment or directly gaging parts.
  • Diameter of 0.3600 inches
  • Length of 2 inches
  • Class X Minus for high precision
  • Tolerance of -0.00004 decimal inch
  • Made from durable tool steel
  • Features a 4 micro coated finish
  • Suitable for calibration of measuring equipment
  • Can be used to gage parts directly
  • Includes certificate of accuracy with NIST traceability