Learn More
MSC Vermont Gage Pin Gage, 0.3865 inch, Class ZZ Minus, Steel
Catalog No. 5000535702
Encompass
This Vermont Gage Class ZZ Minus pin gage is a precision tool designed for accurate measurement and inspection. Constructed from durable steel, it features a 0.3865 inch diameter and a 10 microinch finish for reliable performance in various gaging applications.
  • Economical, general-purpose pin gage for quick and accurate measurements.
  • Suitable for measuring hole sizes, calculating distances between holes, gaging slot widths, checking locations, and Go/NoGo gaging.
  • Offers extended wear life compared to closer tolerance gages.
  • Features a Class ZZ tolerance of 0.0002 "
  • Made from tool steel with a 60/65 Rc hardness.
  • Provides a 10 microinch finish or better.
  • NIST traceable with included Certificate of Accuracy.
Catalog No. 50-005-35702 Supplier MSC Supplier No. 88938659
This Vermont Gage Class ZZ Minus pin gage is a precision tool designed for accurate measurement and inspection. Constructed from durable steel, it features a 0.3865 inch diameter and a 10 microinch finish for reliable performance in various gaging applications.
  • Economical, general-purpose pin gage for quick and accurate measurements.
  • Suitable for measuring hole sizes, calculating distances between holes, gaging slot widths, checking locations, and Go/NoGo gaging.
  • Offers extended wear life compared to closer tolerance gages.
  • Features a Class ZZ tolerance of 0.0002 "
  • Made from tool steel with a 60/65 Rc hardness.
  • Provides a 10 microinch finish or better.
  • NIST traceable with included Certificate of Accuracy.