Learn More
MSC Vermont Gage Pin Gage, 0.3885 in, Class ZZ, Minus, Tool Steel
Catalog No. 5000530450
Encompass
This precision pin gage is designed for accurate measurement in various applications. Crafted from durable tool steel, it features a Class ZZ tolerance for reliable performance in quality control and calibration tasks.
  • Measures hole sizes within a tolerance of +.0000/-.0002 in.
  • Includes a certificate of accuracy with NIST traceability.
  • Constructed from tool steel with a hardness of Rc 60/65.
  • Features a 2 in overall length.
  • Provides a 10 microfinish or better for enhanced precision.
  • Suitable for NoGo inspection and setting various measuring devices.
Catalog No. 50-005-30450 Supplier MSC Supplier No. 89038855
This precision pin gage is designed for accurate measurement in various applications. Crafted from durable tool steel, it features a Class ZZ tolerance for reliable performance in quality control and calibration tasks.
  • Measures hole sizes within a tolerance of +.0000/-.0002 in.
  • Includes a certificate of accuracy with NIST traceability.
  • Constructed from tool steel with a hardness of Rc 60/65.
  • Features a 2 in overall length.
  • Provides a 10 microfinish or better for enhanced precision.
  • Suitable for NoGo inspection and setting various measuring devices.