Learn More
MSC Vermont Gage Pin Gage, 0.4055 in Diameter, Plus, Class ZZ
Catalog No. 5000537210
Encompass
This Vermont Gage pin gage is designed for precise measurement of hole sizes. Made from durable tool steel with a Rc 60/65 hardness and a 10 microfinish, this Class ZZ, Plus (Go) type gage has a diameter of 0.4055 inch and a tolerance of +.0002/-.0000. It includes a Certificate of Accuracy with NIST Traceability to ensure reliable results.
  • Made from tool steel with Rc 60/65 hardness.
  • Features a 10 microfinish or better.
  • Measures hole sizes within a +.0002/-.0000 inch tolerance.
  • Includes a certificate of accuracy with NIST traceability.
  • Has an overall length of 2 inches.
  • Designed as a Go (plus) type gage.
Catalog No. 50-005-37210 Supplier MSC Supplier No. 88940556
This Vermont Gage pin gage is designed for precise measurement of hole sizes. Made from durable tool steel with a Rc 60/65 hardness and a 10 microfinish, this Class ZZ, Plus (Go) type gage has a diameter of 0.4055 inch and a tolerance of +.0002/-.0000. It includes a Certificate of Accuracy with NIST Traceability to ensure reliable results.
  • Made from tool steel with Rc 60/65 hardness.
  • Features a 10 microfinish or better.
  • Measures hole sizes within a +.0002/-.0000 inch tolerance.
  • Includes a certificate of accuracy with NIST traceability.
  • Has an overall length of 2 inches.
  • Designed as a Go (plus) type gage.