Learn More
MSC Vermont Gage Pin Gage, 0.4200 in, Class X Minus, 2 in
Catalog No. 5000531536
Encompass
This pin gage is engineered for precise measurement and inspection tasks. It features a 0.4200 inch diameter, Class X Minus tolerance, and a 2 inch length, complemented by a 4 micro coated finish for enhanced durability and measurement accuracy.
  • Precise 0.4200 inch diameter
  • Class X Minus tolerance
  • 2 inch length
  • 4 micro coated finish
  • Comes with a factory warranty
  • Available from an authorized distributor
  • California Proposition 65 warning
Catalog No. 50-005-31536 Supplier MSC Supplier No. 88842000
This pin gage is engineered for precise measurement and inspection tasks. It features a 0.4200 inch diameter, Class X Minus tolerance, and a 2 inch length, complemented by a 4 micro coated finish for enhanced durability and measurement accuracy.
  • Precise 0.4200 inch diameter
  • Class X Minus tolerance
  • 2 inch length
  • 4 micro coated finish
  • Comes with a factory warranty
  • Available from an authorized distributor
  • California Proposition 65 warning