Learn More
MSC Vermont Gage Pin Gage, 0.4245 in, Class ZZ, No-Go (Minus), Bright Tool Steel
Catalog No. 5000531699
Encompass
This 0.4245 inch Class ZZ No-Go (Minus) Pin Gage is crafted from bright tool steel, ensuring precision and durability for various measurement applications. It is NIST traceable and includes a certificate of accuracy.
  • No-go pin (minus) -.0002 inch / +.0000 inch
  • 2 inch long
  • Tool steel 60/65 HRC
  • 10 microinch finish or better
  • Round within 0.0001 inch or 0.0025 mm
  • Marked with size and direction of tolerance on gages over 0.0605 inch
  • Certificate of accuracy included (printed on the bag the pin is packaged in)
  • Made in U.S.A.
Catalog No. 50-005-31699 Supplier MSC Supplier No. 88942453
This 0.4245 inch Class ZZ No-Go (Minus) Pin Gage is crafted from bright tool steel, ensuring precision and durability for various measurement applications. It is NIST traceable and includes a certificate of accuracy.
  • No-go pin (minus) -.0002 inch / +.0000 inch
  • 2 inch long
  • Tool steel 60/65 HRC
  • 10 microinch finish or better
  • Round within 0.0001 inch or 0.0025 mm
  • Marked with size and direction of tolerance on gages over 0.0605 inch
  • Certificate of accuracy included (printed on the bag the pin is packaged in)
  • Made in U.S.A.