Learn More
MSC Vermont Gage Pin Gage, 0.4635 in, Class ZZ, Plus Tolerance, Tool Steel
Catalog No. 5000531849
Encompass
Vermont Gage Class ZZ Plus Pin Gages are economical, general-purpose tools designed for quick and accurate measurements. They are suitable for measuring hole sizes, calculating distances between holes, gaging slot widths, and checking locations. These gages can be used for Go/NoGo gaging and sizing in micrometers and snap gages, offering added wear life compared to closer tolerance gages.
  • Made from tool steel with a hardness of 60-65 Rc.
  • Has an overall length of 2 inches.
  • Features Class ZZ tolerance, Go (Plus) type.
  • Provides a finish of 10 microinch or better.
  • Roundness is maintained within .0001 "
  • Includes a certificate of accuracy with NIST traceability.
  • Marked with size and direction of tolerance for gages over .0605 "
Catalog No. 50-005-31849 Supplier MSC Supplier No. 88946355
Vermont Gage Class ZZ Plus Pin Gages are economical, general-purpose tools designed for quick and accurate measurements. They are suitable for measuring hole sizes, calculating distances between holes, gaging slot widths, and checking locations. These gages can be used for Go/NoGo gaging and sizing in micrometers and snap gages, offering added wear life compared to closer tolerance gages.
  • Made from tool steel with a hardness of 60-65 Rc.
  • Has an overall length of 2 inches.
  • Features Class ZZ tolerance, Go (Plus) type.
  • Provides a finish of 10 microinch or better.
  • Roundness is maintained within .0001 "
  • Includes a certificate of accuracy with NIST traceability.
  • Marked with size and direction of tolerance for gages over .0605 "