Learn More
MSC Vermont Gage Pin Gage, 0.4660 Inch, Class ZZ, Minus
Catalog No. 5000512968
Encompass
This Class ZZ Minus pin gage is a precision instrument designed for accurate measurement and inspection. With a 0.4660 inch diameter and 2 inch overall length, this steel gage features a 10 microfinish. It is ideal for NoGo inspection and for setting various measuring devices such as micrometers and verniers.
  • For NoGo inspection
  • Set micrometers, verniers, and other measuring devices
  • Includes certificate of accuracy with NIST traceability
  • Individual gage
  • Made in USA
Catalog No. 50-005-12968 Supplier MSC Supplier No. 89046601
This Class ZZ Minus pin gage is a precision instrument designed for accurate measurement and inspection. With a 0.4660 inch diameter and 2 inch overall length, this steel gage features a 10 microfinish. It is ideal for NoGo inspection and for setting various measuring devices such as micrometers and verniers.
  • For NoGo inspection
  • Set micrometers, verniers, and other measuring devices
  • Includes certificate of accuracy with NIST traceability
  • Individual gage
  • Made in USA