Learn More
MSC Vermont Gage Pin Gage, 0.4905 Inch, Class ZZ Minus, Steel
Catalog No. 5000531650
Encompass
This pin gage is designed for precise measurement applications, including setting micrometers, verniers, and other measuring devices, as well as for NoGo inspection. It features a 0.4905 inch diameter and is constructed from durable steel.
  • Class ZZ for precise measurements.
  • 0.4905 inch diameter.
  • Constructed from steel for durability.
  • Suitable for NoGo inspection.
  • Ideal for setting micrometers, verniers, and other measuring devices.
  • Tolerance of 0.0002 "
  • Features a minus direction of tolerance.
  • Reversible style.
Catalog No. 50-005-31650 Supplier MSC Supplier No. 89049050
This pin gage is designed for precise measurement applications, including setting micrometers, verniers, and other measuring devices, as well as for NoGo inspection. It features a 0.4905 inch diameter and is constructed from durable steel.
  • Class ZZ for precise measurements.
  • 0.4905 inch diameter.
  • Constructed from steel for durability.
  • Suitable for NoGo inspection.
  • Ideal for setting micrometers, verniers, and other measuring devices.
  • Tolerance of 0.0002 "
  • Features a minus direction of tolerance.
  • Reversible style.