Learn More
MSC Vermont Gage Pin Gage, 0.493 In, Class ZZ, Minus, Black Oxide
Catalog No. 5000531219
Encompass
This Class ZZ No-Go Pin Gage features a 0.4930 in diameter and a black oxide finish, designed for precise measurements. Constructed from steel, it boasts an overall length of 2 in and meets ISO 17025 standards. It includes a Certificate of Accuracy with NIST Traceability, ensuring reliable performance in measuring hole sizes.
  • Made of steel
  • Tolerance of 0.00020 in
  • Measures hole sizes within +0.0000/-0.0002 in
  • Includes certificate of accuracy with NIST traceability
  • Overall length of 2 in
  • Hardness RC 60/65
  • Surface finish 10 microinch or better
  • Meets ISO 17025 standards
Catalog No. 50-005-31219 Supplier MSC Supplier No. 86562329
This Class ZZ No-Go Pin Gage features a 0.4930 in diameter and a black oxide finish, designed for precise measurements. Constructed from steel, it boasts an overall length of 2 in and meets ISO 17025 standards. It includes a Certificate of Accuracy with NIST Traceability, ensuring reliable performance in measuring hole sizes.
  • Made of steel
  • Tolerance of 0.00020 in
  • Measures hole sizes within +0.0000/-0.0002 in
  • Includes certificate of accuracy with NIST traceability
  • Overall length of 2 in
  • Hardness RC 60/65
  • Surface finish 10 microinch or better
  • Meets ISO 17025 standards