Learn More
MSC Vermont Gage Pin Gage, 0.57 mm Diameter, 2 in OAL, Class ZZ Minus
Catalog No. 5000512316
Encompass
This Vermont Gage Class ZZ Minus pin gage is engineered for accurate measurement and inspection, ideal for calibrating micrometers, performing NoGo inspections, and use with verniers and other measuring devices. Constructed from durable tool steel with a 10 microfinish, it ensures reliable performance.
  • Precise 0.57 mm diameter
  • Overall length of 2 inches
  • Designed for various measurement and inspection applications
  • Made from robust tool steel
  • Features a high-quality 10 microfinish or better
  • Includes a certificate of accuracy with NIST traceability
  • Manufactured in the United States
Catalog No. 50-005-12316 Supplier MSC Supplier No. 40428021
This Vermont Gage Class ZZ Minus pin gage is engineered for accurate measurement and inspection, ideal for calibrating micrometers, performing NoGo inspections, and use with verniers and other measuring devices. Constructed from durable tool steel with a 10 microfinish, it ensures reliable performance.
  • Precise 0.57 mm diameter
  • Overall length of 2 inches
  • Designed for various measurement and inspection applications
  • Made from robust tool steel
  • Features a high-quality 10 microfinish or better
  • Includes a certificate of accuracy with NIST traceability
  • Manufactured in the United States