Learn More
MSC Vermont Gage Pin Gage, 0.598 in, Plus Class ZZ, Tool Steel
Catalog No. 5000530878
Encompass
Class ZZ Pin Gages provide economical, accurate measurement of hole diameters. These long-life metal plugs feature a plus tolerance of 0.0000/+0.0002 inches, ideal for small holes. Each gage offers precision, speed, NIST traceability, Certificate of Accuracy, and electro-etched sizing. They are 2-inch long, centerless lapped, and heat-treated to 60-62 RC for corrosion prevention.
  • Precise hole diameter measurement.
  • Fast measurement activities.
  • Electro-etched sizing for easy identification.
  • Economical, long-lasting; resistant to corrosion/wear.
  • Features 0.0000/+0.0002-inch plus tolerance, NIST traceability, and Certificate of Accuracy.
  • Gages are 2-inch long, centerless lapped, heat-treated to 60-62 RC.
  • Well-ground ends, no sharp edges.
  • Available in steel or Black Guard with 10 micro finish.
Catalog No. 50-005-30878 Supplier MSC Supplier No. 88959804
Class ZZ Pin Gages provide economical, accurate measurement of hole diameters. These long-life metal plugs feature a plus tolerance of 0.0000/+0.0002 inches, ideal for small holes. Each gage offers precision, speed, NIST traceability, Certificate of Accuracy, and electro-etched sizing. They are 2-inch long, centerless lapped, and heat-treated to 60-62 RC for corrosion prevention.
  • Precise hole diameter measurement.
  • Fast measurement activities.
  • Electro-etched sizing for easy identification.
  • Economical, long-lasting; resistant to corrosion/wear.
  • Features 0.0000/+0.0002-inch plus tolerance, NIST traceability, and Certificate of Accuracy.
  • Gages are 2-inch long, centerless lapped, heat-treated to 60-62 RC.
  • Well-ground ends, no sharp edges.
  • Available in steel or Black Guard with 10 micro finish.