Learn More
MSC Vermont Gage Pin Gage, 0.6260 in Dia, 2 in L, Class X Minus
Catalog No. 5000530846
Encompass
This Vermont Gage pin gage is a precision measuring tool designed for accurate calibration and direct gaging of parts. It features a 0.6260 in diameter and a 2 in length, with a Class X Minus tolerance for high accuracy. Manufactured from durable tool steel, it also boasts a 4 micro coated finish.
  • Features a durable tool steel construction.
  • Provides high accuracy with a Class X Minus tolerance.
  • Includes a certificate of accuracy with NIST traceability.
  • Suitable for calibrating measuring equipment or direct gaging of parts.
  • Equipped with a 4 micro coated finish.
Catalog No. 50-005-30846 Supplier MSC Supplier No. 88862602
This Vermont Gage pin gage is a precision measuring tool designed for accurate calibration and direct gaging of parts. It features a 0.6260 in diameter and a 2 in length, with a Class X Minus tolerance for high accuracy. Manufactured from durable tool steel, it also boasts a 4 micro coated finish.
  • Features a durable tool steel construction.
  • Provides high accuracy with a Class X Minus tolerance.
  • Includes a certificate of accuracy with NIST traceability.
  • Suitable for calibrating measuring equipment or direct gaging of parts.
  • Equipped with a 4 micro coated finish.