Learn More
MSC Vermont Gage Pin Gage, 0.7420 in Diameter, Class ZZ Minus, Steel
Catalog No. 5000536871
Encompass
This pin gage is designed for highly accurate measurements in various industrial and laboratory applications. It is particularly suitable for setting micrometers, conducting no-go inspections, and verifying measurements with verniers and other devices. The gage ensures precision with its specific tolerance and durable construction.
  • Made from tool steel with 60/65 Rc hardness.
  • Features a 10 microinch finish or better.
  • Overall length of 2 in.
  • Round within 0.0001 in.
  • Includes a certificate of accuracy with NIST traceability.
  • Suitable for go/no-go gaging and measuring hole sizes.
  • Can be used to calculate distances between holes and gage slot widths.
  • Also used for checking locations and sizing in micrometers and snap gages.
Catalog No. 50-005-36871 Supplier MSC Supplier No. 89074207
This pin gage is designed for highly accurate measurements in various industrial and laboratory applications. It is particularly suitable for setting micrometers, conducting no-go inspections, and verifying measurements with verniers and other devices. The gage ensures precision with its specific tolerance and durable construction.
  • Made from tool steel with 60/65 Rc hardness.
  • Features a 10 microinch finish or better.
  • Overall length of 2 in.
  • Round within 0.0001 in.
  • Includes a certificate of accuracy with NIST traceability.
  • Suitable for go/no-go gaging and measuring hole sizes.
  • Can be used to calculate distances between holes and gage slot widths.
  • Also used for checking locations and sizing in micrometers and snap gages.