Learn More
MSC Vermont Gage Pin Gage, 0.8355 Inch, Plus, Class ZZ, Tool Steel
Catalog No. 5000533297
Encompass
This pin gage provides quick and accurate measurements for various applications, including measuring hole diameters, gaging slot widths, and Go/NoGo inspection. It can also be used to set micrometers and other measuring devices.
  • Tolerance: Class ZZ (0.0002 in or 0.005 mm tolerance)
  • Go (Plus) designation for precise gaging
  • Made from durable tool steel with a hardness of 60/65 Rc
  • Measures 2 in long with a 0.8355 in gage size
  • Features a 10 microinch finish or better
  • Round within 0.0001 in or 0.0025 mm
  • Gages over 0.0605 in are marked with diameter and direction of tolerance
  • Includes a certificate of accuracy
  • NIST traceable for reliable measurements
Catalog No. 50-005-33297 Supplier MSC Supplier No. 06919377
This pin gage provides quick and accurate measurements for various applications, including measuring hole diameters, gaging slot widths, and Go/NoGo inspection. It can also be used to set micrometers and other measuring devices.
  • Tolerance: Class ZZ (0.0002 in or 0.005 mm tolerance)
  • Go (Plus) designation for precise gaging
  • Made from durable tool steel with a hardness of 60/65 Rc
  • Measures 2 in long with a 0.8355 in gage size
  • Features a 10 microinch finish or better
  • Round within 0.0001 in or 0.0025 mm
  • Gages over 0.0605 in are marked with diameter and direction of tolerance
  • Includes a certificate of accuracy
  • NIST traceable for reliable measurements