Learn More
MSC Vermont Gage Pin Gage, 0.8590 in, Class ZZ, Minus, Steel
Catalog No. 5000531115
Encompass
This pin gage is designed for precise measurement applications, including inspecting hole diameters, gaging slot widths, and performing Go/NoGo inspections. It can also be used for setting micrometers, verniers, and other measuring devices. Crafted from steel, this gage features a 0.8590 in diameter and a Class ZZ Minus tolerance, ensuring high accuracy for critical measurements. It measures 2 in long and includes a Certificate of Accuracy with NIST Traceability.
  • Class ZZ tolerance for high accuracy
  • NoGo (Minus) inspection
  • Tool steel construction; 60/65 Rc
  • 2 in length
  • 10 microinch finish or better
  • Round within 0.0001 in
  • Marked with diameter and direction of tolerance
  • Certificate of accuracy with NIST traceability included
Catalog No. 50-005-31115 Supplier MSC Supplier No. 89085906
This pin gage is designed for precise measurement applications, including inspecting hole diameters, gaging slot widths, and performing Go/NoGo inspections. It can also be used for setting micrometers, verniers, and other measuring devices. Crafted from steel, this gage features a 0.8590 in diameter and a Class ZZ Minus tolerance, ensuring high accuracy for critical measurements. It measures 2 in long and includes a Certificate of Accuracy with NIST Traceability.
  • Class ZZ tolerance for high accuracy
  • NoGo (Minus) inspection
  • Tool steel construction; 60/65 Rc
  • 2 in length
  • 10 microinch finish or better
  • Round within 0.0001 in
  • Marked with diameter and direction of tolerance
  • Certificate of accuracy with NIST traceability included