Learn More
MSC Vermont Gage Pin Gage, .0945 in, Class ZZ, Minus Tolerance, Steel
Catalog No. 5000537179
Encompass
This Class ZZ Pin Gage is designed for quick and accurate measurements in various applications, including gaging hole diameters, calculating distances between holes, inspecting slot widths and locations, and performing Go/NoGo checks. It can also be used to precisely set micrometers, verniers, and other measuring devices.
  • Class ZZ tolerance of .0002 in or 0.005 mm
  • Manufactured from tool steel with a hardness of 60/65 Rc
  • Features a 10 microinch finish or better
  • Round within .0001 in or 0.0025 mm
  • Gage length is 2 in
  • Certificate of Accuracy included
  • ISO 17025 accredited and non-accredited calibration certificates may be purchased
Catalog No. 50-005-37179 Supplier MSC Supplier No. 89009450
This Class ZZ Pin Gage is designed for quick and accurate measurements in various applications, including gaging hole diameters, calculating distances between holes, inspecting slot widths and locations, and performing Go/NoGo checks. It can also be used to precisely set micrometers, verniers, and other measuring devices.
  • Class ZZ tolerance of .0002 in or 0.005 mm
  • Manufactured from tool steel with a hardness of 60/65 Rc
  • Features a 10 microinch finish or better
  • Round within .0001 in or 0.0025 mm
  • Gage length is 2 in
  • Certificate of Accuracy included
  • ISO 17025 accredited and non-accredited calibration certificates may be purchased