Learn More
MSC Vermont Gage Pin Gage, 12.59 mm Diameter, Steel, Class ZZ Minus
Catalog No. 5000512052
Encompass
This Class ZZ Minus Plug & Pin Gage is a precision measuring tool designed for no-go inspection and setting various measuring devices such as micrometers and verniers. It features a 12.59 mm diameter and a tight 0.0050 mm tolerance, crafted from durable steel with a 10 microfinish.
  • Designed for no-go inspection
  • Can set micrometers, verniers, and other measuring devices
  • Includes certificate of accuracy with NIST traceability
  • Made from steel
  • Features a 10 microfinish
Catalog No. 50-005-12052 Supplier MSC Supplier No. 40440042
This Class ZZ Minus Plug & Pin Gage is a precision measuring tool designed for no-go inspection and setting various measuring devices such as micrometers and verniers. It features a 12.59 mm diameter and a tight 0.0050 mm tolerance, crafted from durable steel with a 10 microfinish.
  • Designed for no-go inspection
  • Can set micrometers, verniers, and other measuring devices
  • Includes certificate of accuracy with NIST traceability
  • Made from steel
  • Features a 10 microfinish