Learn More
MSC Vermont Gage Pin Gage, .1375 in, Class X, Go (Plus), 2 in
Catalog No. 5000530927
Encompass
This Class X Go (Plus) pin gage is designed for precise measurement applications. It serves as an accurate, cost-effective standard for calibrating measuring equipment or directly gaging parts, suitable for both shop floor and metrology laboratory environments.
  • Class X tolerance
  • NIST traceable
  • Constructed from tool steel, 60/65 Rc (cold stabilized)
  • 2 inch long
  • Features a 4 microinch finish or better
  • Round within 1/2 of tolerance
  • Certificate of accuracy included
Catalog No. 50-005-30927 Supplier MSC Supplier No. 88713755
This Class X Go (Plus) pin gage is designed for precise measurement applications. It serves as an accurate, cost-effective standard for calibrating measuring equipment or directly gaging parts, suitable for both shop floor and metrology laboratory environments.
  • Class X tolerance
  • NIST traceable
  • Constructed from tool steel, 60/65 Rc (cold stabilized)
  • 2 inch long
  • Features a 4 microinch finish or better
  • Round within 1/2 of tolerance
  • Certificate of accuracy included