Learn More
MSC Vermont Gage Pin Gage, 14.40 mm, Minus Class ZZ, Steel
Catalog No. 5000527903
Encompass
This pin gage is made from steel and is designed for precise measurement with a 14.40 mm size and minus Class ZZ tolerance. Its reversible style provides versatility for various applications.
  • Standard ISO 17025 accredited calibration option available
  • Expedited ISO 17025 accredited calibration option available
  • Custom laser engraving for gage ID or other naming conventions
  • Standard calibration offers a 5-day turnaround
  • Expedited calibration completed in 2-3 business days
  • Certificates are stored on the cloud-based A-Trax system
  • Constructed from steel
  • Reversible design
  • Measurements in millimeters
  • Features Class ZZ tolerance
  • Minus tolerance direction
Catalog No. 50-005-27903 Supplier MSC Supplier No. 40441859
This pin gage is made from steel and is designed for precise measurement with a 14.40 mm size and minus Class ZZ tolerance. Its reversible style provides versatility for various applications.
  • Standard ISO 17025 accredited calibration option available
  • Expedited ISO 17025 accredited calibration option available
  • Custom laser engraving for gage ID or other naming conventions
  • Standard calibration offers a 5-day turnaround
  • Expedited calibration completed in 2-3 business days
  • Certificates are stored on the cloud-based A-Trax system
  • Constructed from steel
  • Reversible design
  • Measurements in millimeters
  • Features Class ZZ tolerance
  • Minus tolerance direction