Learn More
MSC Vermont Gage Pin Gage, 14.59 mm, Class ZZ, Tool Steel
Catalog No. 5000528895
Encompass
This Vermont Gage pin gage is designed for precise 'Go' inspection and is also suitable for setting micrometers, verniers, and other measuring devices. It features a reversible style and is constructed from durable tool steel.
  • Designed for go inspection
  • Used to set micrometers, verniers, and other measuring devices
  • Class ZZ precision
  • Diameter of 14.59 mm
  • Plus direction of tolerance
  • Constructed from tool steel
  • Reversible style
  • Metric measurement system
  • Tolerance of 0.00508 mm
Catalog No. 50-005-28895 Supplier MSC Supplier No. 68105642
This Vermont Gage pin gage is designed for precise 'Go' inspection and is also suitable for setting micrometers, verniers, and other measuring devices. It features a reversible style and is constructed from durable tool steel.
  • Designed for go inspection
  • Used to set micrometers, verniers, and other measuring devices
  • Class ZZ precision
  • Diameter of 14.59 mm
  • Plus direction of tolerance
  • Constructed from tool steel
  • Reversible style
  • Metric measurement system
  • Tolerance of 0.00508 mm