Learn More
MSC Vermont Gage Pin Gage, 14.67 mm, Steel, Class ZZ, Go (Plus)
Catalog No. 5000527677
Encompass
Make quick and accurate measurements with these pin gages, ideal for checking hole sizes, calculating distances between holes, gaging slot widths, and verifying locations. They can also be used for Go/NoGo gaging and sizing in micrometers and snap gages.
  • Tool steel: 60/65 Rc
  • 2 inches long
  • 10 microinch finish or better
  • Round within .0001 inches
  • Marked with size and direction of tolerance on gages over .0605 inches
Catalog No. 50-005-27677 Supplier MSC Supplier No. 68105725
Make quick and accurate measurements with these pin gages, ideal for checking hole sizes, calculating distances between holes, gaging slot widths, and verifying locations. They can also be used for Go/NoGo gaging and sizing in micrometers and snap gages.
  • Tool steel: 60/65 Rc
  • 2 inches long
  • 10 microinch finish or better
  • Round within .0001 inches
  • Marked with size and direction of tolerance on gages over .0605 inches