Learn More
MSC Vermont Gage Pin Gage, 15.01 mm, Steel, Class ZZ, Go (Plus)
Catalog No. 5000514705
Encompass
This pin gage is designed for quick and accurate measurements in various applications. It can be used to measure hole sizes, calculate distances between holes, gage slot widths, and check locations. This gage is also suitable for Go/NoGo gaging and sizing in micrometers and snap gages.
  • Tool steel construction with 60/65 Rc hardness.
  • Measures 2 in long.
  • Features a 10 microinch finish or better.
  • Guaranteed round within .0001 in.
  • Marked with size and direction of tolerance for gages over .0605 in.
Catalog No. 50-005-14705 Supplier MSC Supplier No. 68106061
This pin gage is designed for quick and accurate measurements in various applications. It can be used to measure hole sizes, calculate distances between holes, gage slot widths, and check locations. This gage is also suitable for Go/NoGo gaging and sizing in micrometers and snap gages.
  • Tool steel construction with 60/65 Rc hardness.
  • Measures 2 in long.
  • Features a 10 microinch finish or better.
  • Guaranteed round within .0001 in.
  • Marked with size and direction of tolerance for gages over .0605 in.