Learn More
MSC Vermont Gage Pin Gage, 15.60 mm, Class ZZ, Minus Tolerance
Catalog No. 5000532850
Encompass
This pin gage is designed for precise measurements in various industrial and laboratory applications. Manufactured from tool steel, it features a 15.60 mm diameter with a Class ZZ minus tolerance, ensuring high accuracy for gaging and setting measuring devices. Each gage includes a Certificate of Accuracy with NIST traceability.
  • Material is tool steel; 60/65 Rc
  • Finish is 10 microinch or better
  • Overall length is 2 in
  • Features Class ZZ tolerance (.0002" or .005mm)
  • Includes Certificate of Accuracy with NIST traceability
  • Measures hole sizes, gaging slot widths, and checks locations
  • Suitable for setting micrometers, verniers, and other measuring devices
Catalog No. 50-005-32850 Supplier MSC Supplier No. 40443053
This pin gage is designed for precise measurements in various industrial and laboratory applications. Manufactured from tool steel, it features a 15.60 mm diameter with a Class ZZ minus tolerance, ensuring high accuracy for gaging and setting measuring devices. Each gage includes a Certificate of Accuracy with NIST traceability.
  • Material is tool steel; 60/65 Rc
  • Finish is 10 microinch or better
  • Overall length is 2 in
  • Features Class ZZ tolerance (.0002" or .005mm)
  • Includes Certificate of Accuracy with NIST traceability
  • Measures hole sizes, gaging slot widths, and checks locations
  • Suitable for setting micrometers, verniers, and other measuring devices