Learn More
MSC Vermont Gage Pin Gage, 15.67 mm, Class ZZ, Go (Plus)
Catalog No. 5000512695
Encompass
This pin gage is a Go (Plus) type, featuring a diameter of 15.67 mm and an overall length of 2 in. It is constructed from tool steel with a hardness of Rc 60/65 and a surface finish of steel - 10 microfinish or better. The gage has a tolerance of +.005/-.000, making it suitable for measuring hole sizes within this specification. This product is designated as Class ZZ and is manufactured in the United States.
  • Constructed from tool steel; Rc 60/65
  • Tolerance of +.005/-.000
  • Overall length of 2 in
  • Finish of steel - 10 microfinish or better
  • Measures hole sizes within +.005/-.000"
  • Includes certificate of accuracy with NIST traceability
Catalog No. 50-005-12695 Supplier MSC Supplier No. 68106723
This pin gage is a Go (Plus) type, featuring a diameter of 15.67 mm and an overall length of 2 in. It is constructed from tool steel with a hardness of Rc 60/65 and a surface finish of steel - 10 microfinish or better. The gage has a tolerance of +.005/-.000, making it suitable for measuring hole sizes within this specification. This product is designated as Class ZZ and is manufactured in the United States.
  • Constructed from tool steel; Rc 60/65
  • Tolerance of +.005/-.000
  • Overall length of 2 in
  • Finish of steel - 10 microfinish or better
  • Measures hole sizes within +.005/-.000"
  • Includes certificate of accuracy with NIST traceability