Learn More
MSC Vermont Gage Pin Gage, 15.90 mm Diameter, Steel, Class ZZ, NoGo (Minus)
Catalog No. 5000529551
Encompass
These pin gages allow for quick and accurate measurements. They are ideal for determining hole sizes, calculating distances between holes, gaging slot widths, checking locations, and for use in Go/NoGo applications, as well as sizing with micrometers and snap gages.
  • Tool steel construction with 60/65 Rc hardness
  • 2 in length
  • Features a 10 microinch finish or better
  • Round within 0.0001 in
  • Marked with size and direction of tolerance for gages over 0.0605 in
Catalog No. 50-005-29551 Supplier MSC Supplier No. 40443350
These pin gages allow for quick and accurate measurements. They are ideal for determining hole sizes, calculating distances between holes, gaging slot widths, checking locations, and for use in Go/NoGo applications, as well as sizing with micrometers and snap gages.
  • Tool steel construction with 60/65 Rc hardness
  • 2 in length
  • Features a 10 microinch finish or better
  • Round within 0.0001 in
  • Marked with size and direction of tolerance for gages over 0.0605 in