Learn More
MSC Vermont Gage Pin Gage, 18.24 mm, Class ZZ Plus, Tool Steel
Catalog No. 5000531857
Encompass
This pin gage measures hole sizes with precision. It features durable tool steel construction (Rc 60/65) for consistent performance. With an 18.24 mm diameter and Plus Class ZZ tolerance (+.005/-.000), it includes a Certificate of Accuracy with NIST Traceability. The 10 microfinish or better steel finish and 2 in overall length make it a reliable tool for exacting measurement applications.
  • Durable tool steel construction (Rc 60/65).
  • Tolerance of +.005/-.000 for precise hole size measurement.
  • Includes a certificate of accuracy with NIST traceability.
  • Overall length of 2 in.
  • 10 microfinish or better steel finish.
Catalog No. 50-005-31857 Supplier MSC Supplier No. 68109297
This pin gage measures hole sizes with precision. It features durable tool steel construction (Rc 60/65) for consistent performance. With an 18.24 mm diameter and Plus Class ZZ tolerance (+.005/-.000), it includes a Certificate of Accuracy with NIST Traceability. The 10 microfinish or better steel finish and 2 in overall length make it a reliable tool for exacting measurement applications.
  • Durable tool steel construction (Rc 60/65).
  • Tolerance of +.005/-.000 for precise hole size measurement.
  • Includes a certificate of accuracy with NIST traceability.
  • Overall length of 2 in.
  • 10 microfinish or better steel finish.