Learn More
MSC Vermont Gage Pin Gage, 18.39 mm, Class ZZ, Minus, Steel
Catalog No. 5000529053
Encompass
This metric pin gage is designed for precise measurement applications, ensuring accurate inspection of bore diameters. Constructed from durable steel, it offers reliability and longevity. Its reversible style enhances usability and extends its service life, making it an essential tool for quality control and manufacturing processes that require exact measurements.
  • Metric system of measurement
  • Class ZZ tolerance
  • Minus direction of tolerance
  • Steel material ensures durability
  • Reversible style extends service life
  • Ideal for NoGo inspection and setting various measuring devices
  • Includes certificate of accuracy with NIST traceability
  • Features a 10 microinch finish
  • Overall length of 2 inches (50.8 mm)
Catalog No. 50-005-29053 Supplier MSC Supplier No. 40445843
This metric pin gage is designed for precise measurement applications, ensuring accurate inspection of bore diameters. Constructed from durable steel, it offers reliability and longevity. Its reversible style enhances usability and extends its service life, making it an essential tool for quality control and manufacturing processes that require exact measurements.
  • Metric system of measurement
  • Class ZZ tolerance
  • Minus direction of tolerance
  • Steel material ensures durability
  • Reversible style extends service life
  • Ideal for NoGo inspection and setting various measuring devices
  • Includes certificate of accuracy with NIST traceability
  • Features a 10 microinch finish
  • Overall length of 2 inches (50.8 mm)