Learn More
MSC Vermont Gage Pin Gage, 19.26 mm, Class ZZ, Minus Tolerance, Steel
Catalog No. 5000528641
Encompass
This pin gage is designed for precise measurement applications, including setting micrometers, verniers, and other measuring devices, and for NoGo inspection. It features a 19.26 mm diameter and a Class ZZ minus tolerance, providing accuracy for various industrial and laboratory tasks.
  • Class ZZ tolerance
  • NoGo inspection capability
  • Constructed from durable steel
  • Reversible style for extended use
Catalog No. 50-005-28641 Supplier MSC Supplier No. 40446718
This pin gage is designed for precise measurement applications, including setting micrometers, verniers, and other measuring devices, and for NoGo inspection. It features a 19.26 mm diameter and a Class ZZ minus tolerance, providing accuracy for various industrial and laboratory tasks.
  • Class ZZ tolerance
  • NoGo inspection capability
  • Constructed from durable steel
  • Reversible style for extended use