Learn More
MSC Vermont Gage Pin Gage, 19.72 mm, Class ZZ, Go (Plus)
Catalog No. 5000535868
Encompass
This pin gage is engineered for precise measurement applications. Constructed from durable tool steel with a fine microfinish, it is ideal for accurately checking hole sizes.
  • Constructed from tool steel (Rc 60/65)
  • Features a steel microfinish of 10 or better
  • Measures 19.72 mm in diameter
  • Offers a tolerance of +.005/-.000
  • Has an overall length of 2 in
  • Class ZZ accuracy for demanding tasks
  • Go (Plus) type for specific gaging needs
  • Includes certificate of accuracy with NIST traceability
Catalog No. 50-005-35868 Supplier MSC Supplier No. 68110774
This pin gage is engineered for precise measurement applications. Constructed from durable tool steel with a fine microfinish, it is ideal for accurately checking hole sizes.
  • Constructed from tool steel (Rc 60/65)
  • Features a steel microfinish of 10 or better
  • Measures 19.72 mm in diameter
  • Offers a tolerance of +.005/-.000
  • Has an overall length of 2 in
  • Class ZZ accuracy for demanding tasks
  • Go (Plus) type for specific gaging needs
  • Includes certificate of accuracy with NIST traceability