Learn More
MSC Vermont Gage Pin Gage, 19.96 mm, Plus, Class ZZ, Tool Steel
Catalog No. 5000531354
Encompass
This pin gage is engineered for precise measurement of hole sizes, featuring a 19.96 mm diameter and Class ZZ accuracy. Constructed from durable tool steel with a fine finish, it ensures reliable performance in demanding applications and comes with a certificate of accuracy.
  • Tool steel construction with Rc 60/65 hardness
  • Steel with 10 microfinish or better
  • 19.96 mm diameter
  • Go (plus) type
  • Class ZZ accuracy
  • Tolerance of +.005/-.000
  • 2 in overall length
  • Measures hole sizes accurately
  • Includes certificate of accuracy with NIST traceability
Catalog No. 50-005-31354 Supplier MSC Supplier No. 68111012
This pin gage is engineered for precise measurement of hole sizes, featuring a 19.96 mm diameter and Class ZZ accuracy. Constructed from durable tool steel with a fine finish, it ensures reliable performance in demanding applications and comes with a certificate of accuracy.
  • Tool steel construction with Rc 60/65 hardness
  • Steel with 10 microfinish or better
  • 19.96 mm diameter
  • Go (plus) type
  • Class ZZ accuracy
  • Tolerance of +.005/-.000
  • 2 in overall length
  • Measures hole sizes accurately
  • Includes certificate of accuracy with NIST traceability