Learn More
MSC Vermont Gage Pin Gage, 20.22mm, Class ZZ, Minus
Catalog No. 5000529309
Encompass
This pin gage is manufactured from tool steel with a hardness of Rc 60/65 and a steel - 10 microfinish or better. It features a 20.22 mm diameter and a tolerance of +.000/-.005 in. The overall length is 2 in. It is designed for measuring hole sizes and includes a certificate of accuracy with NIST traceability.
  • Made from tool steel with a hardness of Rc 60/65
  • Provides a tolerance of +.000/-.005 in
  • Measures hole sizes within +.000/-.005 inches
  • Includes a certificate of accuracy with NIST traceability
  • Features an overall length of 2 in
  • Designed as a no-go (minus) pin gage
  • Class ZZ
  • Finished with steel - 10 microfinish or better
  • Diameter of 20.22 mm
Catalog No. 50-005-29309 Supplier MSC Supplier No. 40447674
This pin gage is manufactured from tool steel with a hardness of Rc 60/65 and a steel - 10 microfinish or better. It features a 20.22 mm diameter and a tolerance of +.000/-.005 in. The overall length is 2 in. It is designed for measuring hole sizes and includes a certificate of accuracy with NIST traceability.
  • Made from tool steel with a hardness of Rc 60/65
  • Provides a tolerance of +.000/-.005 in
  • Measures hole sizes within +.000/-.005 inches
  • Includes a certificate of accuracy with NIST traceability
  • Features an overall length of 2 in
  • Designed as a no-go (minus) pin gage
  • Class ZZ
  • Finished with steel - 10 microfinish or better
  • Diameter of 20.22 mm