Learn More
MSC Vermont Gage Pin Gage, 20.66mm, Steel, Class ZZ, NoGo (Minus)
Catalog No. 5000528534
Encompass
This pin gage is designed for precise measurement applications, featuring a 20.66mm diameter and constructed from durable steel. It is a Class ZZ gage with a NoGo (Minus) tolerance, ensuring high accuracy for quality control and inspection tasks.
  • Tool steel construction ensures durability and longevity.
  • Overall length of 2 inches provides ease of handling.
  • Features a 10 microinch finish or better for smooth operation.
  • Round within .0001" for consistent and accurate readings.
  • Marked with size and direction of tolerance for easy identification.
  • Provides an accuracy of .0002" for reliable measurements.
Catalog No. 50-005-28534 Supplier MSC Supplier No. 40448110
This pin gage is designed for precise measurement applications, featuring a 20.66mm diameter and constructed from durable steel. It is a Class ZZ gage with a NoGo (Minus) tolerance, ensuring high accuracy for quality control and inspection tasks.
  • Tool steel construction ensures durability and longevity.
  • Overall length of 2 inches provides ease of handling.
  • Features a 10 microinch finish or better for smooth operation.
  • Round within .0001" for consistent and accurate readings.
  • Marked with size and direction of tolerance for easy identification.
  • Provides an accuracy of .0002" for reliable measurements.