Learn More
MSC Vermont Gage Pin Gage, 20.72 mm, Class ZZ, Minus
Catalog No. 5000528186
Encompass
Vermont Gage Pin Gages are essential tools for precision measurement in various industrial and scientific applications. This specific pin gage is designed for quick and accurate determination of hole sizes, slot widths, and distances between holes, making it suitable for Go/NoGo gaging and precise sizing with micrometers and snap gages.
  • Measures hole sizes, distances between holes, and slot widths.
  • Ideal for Go/NoGo gaging.
  • Can be used for sizing in micrometers and snap gages.
  • Constructed from durable tool steel with a hardness of 60/65 Rc.
  • Features a finish of 10 microinch or better.
  • Maintains accuracy, round within .0001".
  • Includes a Certificate of Accuracy with NIST Traceability.
  • Pin gage length is 2 in.
  • Features Class ZZ tolerance and NoGo (Minus) type.
Catalog No. 50-005-28186 Supplier MSC Supplier No. 40448177
Vermont Gage Pin Gages are essential tools for precision measurement in various industrial and scientific applications. This specific pin gage is designed for quick and accurate determination of hole sizes, slot widths, and distances between holes, making it suitable for Go/NoGo gaging and precise sizing with micrometers and snap gages.
  • Measures hole sizes, distances between holes, and slot widths.
  • Ideal for Go/NoGo gaging.
  • Can be used for sizing in micrometers and snap gages.
  • Constructed from durable tool steel with a hardness of 60/65 Rc.
  • Features a finish of 10 microinch or better.
  • Maintains accuracy, round within .0001".
  • Includes a Certificate of Accuracy with NIST Traceability.
  • Pin gage length is 2 in.
  • Features Class ZZ tolerance and NoGo (Minus) type.