Learn More
MSC Vermont Gage Pin Gage, .2095" Minus, Class ZZ, Tool Steel, 2 in
Catalog No. 5000531908
Encompass
This .2095 inch Minus Class ZZ pin gage is designed for quick and accurate measurements. It can be used for measuring hole diameters, calculating distances between holes, gaging slot widths and locations, and for Go/NoGo inspection. Additionally, it helps in setting micrometers, verniers, and other measuring devices.
  • Constructed from tool steel with a hardness of 60/65 Rc
  • Features a Class ZZ tolerance of .0002 in
  • Measures 2 in long
  • Surface finish of 10 microinch or better
  • Round within .0001 in
  • Includes a certificate of accuracy
  • Gages over .0605 in are marked with diameter and direction of tolerance
Catalog No. 50-005-31908 Supplier MSC Supplier No. 89020952
This .2095 inch Minus Class ZZ pin gage is designed for quick and accurate measurements. It can be used for measuring hole diameters, calculating distances between holes, gaging slot widths and locations, and for Go/NoGo inspection. Additionally, it helps in setting micrometers, verniers, and other measuring devices.
  • Constructed from tool steel with a hardness of 60/65 Rc
  • Features a Class ZZ tolerance of .0002 in
  • Measures 2 in long
  • Surface finish of 10 microinch or better
  • Round within .0001 in
  • Includes a certificate of accuracy
  • Gages over .0605 in are marked with diameter and direction of tolerance