Learn More
MSC Vermont Gage Pin Gage, 21.21 mm, Class ZZ Minus, 2 inch OAL
Catalog No. 5000533887
Encompass
This precision pin gage is designed for accurate measurement in various applications, including setting micrometers, NoGo inspection, and calibrating other measuring devices. Constructed from durable steel, it features a 21.21 mm diameter and a 2-inch overall length, ensuring reliability and accuracy for critical gaging tasks. Each gage includes a Certificate of Accuracy with NIST Traceability.
  • Measures 21.21 mm diameter with a 2 inch overall length.
  • Class ZZ tolerance provides high precision.
  • Manufactured from steel for durability and longevity.
  • Features a 10 microinch finish for smooth operation.
  • Includes a certificate of accuracy with NIST traceability.
  • Ideal for setting micrometers, verniers, and other measuring devices.
  • Suitable for NoGo inspection, measuring hole sizes, and slot widths.
Catalog No. 50-005-33887 Supplier MSC Supplier No. 40448664
This precision pin gage is designed for accurate measurement in various applications, including setting micrometers, NoGo inspection, and calibrating other measuring devices. Constructed from durable steel, it features a 21.21 mm diameter and a 2-inch overall length, ensuring reliability and accuracy for critical gaging tasks. Each gage includes a Certificate of Accuracy with NIST Traceability.
  • Measures 21.21 mm diameter with a 2 inch overall length.
  • Class ZZ tolerance provides high precision.
  • Manufactured from steel for durability and longevity.
  • Features a 10 microinch finish for smooth operation.
  • Includes a certificate of accuracy with NIST traceability.
  • Ideal for setting micrometers, verniers, and other measuring devices.
  • Suitable for NoGo inspection, measuring hole sizes, and slot widths.