Learn More
MSC Vermont Gage Pin Gage, 21.63 mm, Class ZZ, Minus Tolerance
Catalog No. 5000527146
Encompass
This Vermont Gage pin gage is designed for precise measurements in metric applications. It features a 21.63 mm size with a Class ZZ tolerance and a minus designation, making it suitable for checking bore sizes and hole locations.
  • Individual gauge
  • Metric measurement
  • 21.63 mm size
  • Class ZZ tolerance
  • Minus tolerance (.00024 in or .006 mm)
  • Made in USA
Catalog No. 50-005-27146 Supplier MSC Supplier No. 40449084
This Vermont Gage pin gage is designed for precise measurements in metric applications. It features a 21.63 mm size with a Class ZZ tolerance and a minus designation, making it suitable for checking bore sizes and hole locations.
  • Individual gauge
  • Metric measurement
  • 21.63 mm size
  • Class ZZ tolerance
  • Minus tolerance (.00024 in or .006 mm)
  • Made in USA