Learn More
MSC Vermont Gage Pin Gage, 22.19 mm, Class ZZ, Go (Plus), Tool Steel
Catalog No. 5000534410
Encompass
This pin gage is an economical, general-purpose tool designed for quick and accurate measurements. It is ideal for measuring hole sizes, calculating distances between holes, gaging slot widths, and checking locations. This 22.19 mm diameter, Class ZZ, Go (Plus) pin gage is made from durable tool steel, providing reliable performance and extended wear life in various applications.
  • Measures hole sizes within +.005/-.000".
  • Constructed from tool steel with Rc 60/65 hardness.
  • Features a steel finish with 10 microinch or better.
  • Includes a certificate of accuracy with NIST traceability.
  • Go (Plus) type for efficient gaging.
  • Overall length of 2 in.
  • Class ZZ tolerance provides high precision.
Catalog No. 50-005-34410 Supplier MSC Supplier No. 68113240
This pin gage is an economical, general-purpose tool designed for quick and accurate measurements. It is ideal for measuring hole sizes, calculating distances between holes, gaging slot widths, and checking locations. This 22.19 mm diameter, Class ZZ, Go (Plus) pin gage is made from durable tool steel, providing reliable performance and extended wear life in various applications.
  • Measures hole sizes within +.005/-.000".
  • Constructed from tool steel with Rc 60/65 hardness.
  • Features a steel finish with 10 microinch or better.
  • Includes a certificate of accuracy with NIST traceability.
  • Go (Plus) type for efficient gaging.
  • Overall length of 2 in.
  • Class ZZ tolerance provides high precision.