Learn More
MSC Vermont Gage Pin Gage, .3275 in, Class ZZ, Steel
Catalog No. 5000515786
Encompass
Vermont Gage Class ZZ Pin Gages are economical general-purpose pin gages designed for quick and accurate measurements. They are suitable for measuring hole sizes, calculating distances between holes, gaging slot widths, and checking locations. These gages can be used for Go/NoGo gaging and sizing in micrometers and snap gages, offering added wear life.
  • Tolerance: class ZZ (.0002 in or .005 mm tolerance).
  • Material: tool steel, 60/65 Rc.
  • Length: 2 in long standard.
  • Surface finish: 10 microinch finish or better.
  • Roundness: round within .0001 in or .0025 mm.
  • NIST traceable.
  • Certificate of accuracy included.
  • Economical and versatile for go/nogo gaging.
  • Offers added wear life and durability.
  • Can be used for various measurement tasks including hole sizing, distance calculation, slot width gaging, and location checks.
Catalog No. 50-005-15786 Supplier MSC Supplier No. 88932751
Vermont Gage Class ZZ Pin Gages are economical general-purpose pin gages designed for quick and accurate measurements. They are suitable for measuring hole sizes, calculating distances between holes, gaging slot widths, and checking locations. These gages can be used for Go/NoGo gaging and sizing in micrometers and snap gages, offering added wear life.
  • Tolerance: class ZZ (.0002 in or .005 mm tolerance).
  • Material: tool steel, 60/65 Rc.
  • Length: 2 in long standard.
  • Surface finish: 10 microinch finish or better.
  • Roundness: round within .0001 in or .0025 mm.
  • NIST traceable.
  • Certificate of accuracy included.
  • Economical and versatile for go/nogo gaging.
  • Offers added wear life and durability.
  • Can be used for various measurement tasks including hole sizing, distance calculation, slot width gaging, and location checks.