Learn More
MSC Vermont Gage Pin Gage, 5.45 mm Diameter, Class ZZ, Steel, Plus
Catalog No. 5000527239
Encompass
Vermont Gage Class ZZ Plus Pin Gages are economical and durable tools designed for precise measurements in various applications. They are ideal for quick and accurate assessments of hole sizes, distances, slot widths, and for Go/NoGo gaging, also suitable for use with micrometers and snap gages. Each gage is meticulously crafted for reliability and compliance in professional settings.
  • Economical gages with extended wear life
  • Class ZZ tolerance for precision
  • Constructed from tool steel (60/65 Rc)
  • 2 inch length for versatile use
  • Features a 10 microinch finish or better
  • Round within .0001 inch or .0025 mm
  • NIST traceable for accuracy
  • Marked with size and direction of tolerance on gages over .0605 inch
  • Certificate of accuracy included with each package
  • Individually packaged for protection
Catalog No. 50-005-27239 Supplier MSC Supplier No. 68096502
Vermont Gage Class ZZ Plus Pin Gages are economical and durable tools designed for precise measurements in various applications. They are ideal for quick and accurate assessments of hole sizes, distances, slot widths, and for Go/NoGo gaging, also suitable for use with micrometers and snap gages. Each gage is meticulously crafted for reliability and compliance in professional settings.
  • Economical gages with extended wear life
  • Class ZZ tolerance for precision
  • Constructed from tool steel (60/65 Rc)
  • 2 inch length for versatile use
  • Features a 10 microinch finish or better
  • Round within .0001 inch or .0025 mm
  • NIST traceable for accuracy
  • Marked with size and direction of tolerance on gages over .0605 inch
  • Certificate of accuracy included with each package
  • Individually packaged for protection